Merck
Representativeness of extracts of offset paper packaging and analysis of the main odor-active compounds.
Journal of agricultural and food chemistry (2004-04-15)
Pascale Landy, Sophie Nicklaus, Etienne Sémon, Patrick Mielle, Elisabeth Guichard
ABSTRAKT
Packagings often carry odors due to the support and printing inks. The aim of the investigation was to define a representative solvent-free extract of paper-based packaging materials printed by the offset process, for the identification of the odor-causing volatile compounds. Static headspace and solid-phase microextraction were the two applied extraction methods. Representativeness tests showed that the odor of the PDMS fiber extract gave satisfying odor similarities with the original packaging. The sample incubation was performed at 40 degrees C for 30 min, whereas the extraction time was 3 min at 40 degrees C. Extracts of both the nonprinted and printed papers of different batches were analyzed by gas chromatography-olfactometry. 4-Phenylcyclohexene was identified as the most potent compound contributing to the latex-like odor of the nonprinted paper. Among the 13 major odorants identified by mass spectrometry, 10 were aldehydes and ketones generated by oxidation of the printing ink resins. The ratio of odorants to interferences was too low for a possible detection of the key odorants by nonseparative techniques such as sensor arrays.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, red hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub