Merck
Myoepithelial carcinoma treatment in children: a report from the TREP project.
Pediatric blood & cancer (2013-10-19)
Gianni Bisogno, Arianna Tagarelli, Amalia Schiavetti, Giovanni Scarzello, Andrea Ferrari, Giovanni Cecchetto, Rita Alaggio
ABSTRAKT
Myoepithelial carcinoma (MC) of soft tissues is an aggressive tumor that rarely affects children, for whom no established treatment protocols exist. As part of the TREP (Tumori Rari in Età Pediatrica) project - an Italian network dedicated to children and adolescents with very rare tumors - we present a series of patients with MC, who were treated homogeneously and achieved a satisfactory outcome. From 2005 to 2012, seven patients (age 0.5-9.2 years) with a diagnosis of MC were registered in the TREP study. After one patient treated with ifosfamide, cisplatin, and etoposide showed tumor shrinkage and experienced long-term disease remission, all subsequent patients were treated with the same chemotherapy regimen. All patients also received radiotherapy. Initial surgical management involved a biopsy in three cases and tumor resection in 4. Response to initial chemotherapy was evaluable in four patients: two had a partial remission, one a minor response and one stable disease. Four patients received external-beam radiotherapy and three had brachytherapy. Overall, six patients are alive in first complete remission with a median follow-up of 2.5 years (0.9-5.1 years). Though our experience is limited to a small number of patients, our treatment strategy for patients with MC is appears clinically useful and demonstrates how cooperation within the TREP project has enabled enough data to be collected to propose treatment recommendations for pediatric patients with this very rare tumor.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ifosfamide, ≥98%
Ifosfamide, European Pharmacopoeia (EP) Reference Standard
Etoposide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder