Merck
  • Fibrinogen recovery and changes in fibrin-based clot firmness after cryoprecipitate administration in patients undergoing aortic surgery involving deep hypothermic circulatory arrest.
Fibrinogen recovery and changes in fibrin-based clot firmness after cryoprecipitate administration in patients undergoing aortic surgery involving deep hypothermic circulatory arrest.
Transfusion (2013-11-07)
Sang Hyun Lee, Sangmin M Lee, Chung Su Kim, Hyun Sung Cho, Jong-Hwan Lee, Cheol Hee Lee, Eunhee Kim, Kiick Sung, Cristina Solomon, Jingu Kang, Young Ri Kim
ABSTRAKT
Cryoprecipitate may be used to treat bleeding in cardiac surgery. Its effects on plasma fibrinogen and fibrin clotting in this setting are poorly defined. Patients undergoing on-pump aortic surgery with deep hypothermic circulatory arrest (DHCA) were recruited prospectively. After protamine reversal, cryoprecipitate was administered to patients with bleeding, and fibrin deficit was indicated by thromboelastometry (ROTEM)-based FIBTEM test. Coagulation was assessed using ROTEM-based tests and standard laboratory tests before and after cryoprecipitate. Thirteen patients were included. Cryoprecipitate significantly elevated EXTEM A10 from (mean ± standard deviation) 29.4 ± 5.8 to 34.8 ± 5.9 mm (p = 0.01), FIBTEM A10 from 3.5 ± 0.9 to 5.8 ± 1.7 mm (p = 0.04), and plasma fibrinogen concentration from 154.2 ± 25.6 to 193.4 ± 30.5 mg/dL (p = 0.01). EXTEM clot elasticity at 10 minutes (CE10) increased from 42.5 ± 12.0 to 54.7 ± 14.9 mm after cryoprecipitate (30.0% increase). FIBTEM CE10 increased from 3.7 ± 0.9 to 6.2 ± 2.0 mm (53.0% increase). A fibrinogen dose of 13.2 ± 5.2 mg/kg was required to increase FIBTEM A10 by 1 mm. In vivo recovery of fibrinogen was 61.6 ± 31.2%. Cryoprecipitate increased plasma fibrinogen levels and fibrin-based clotting in bleeding patients undergoing aortic surgery with DHCA. In vivo recovery of fibrinogen was considerably below 100% and fibrinogen content varied between cryoprecipitate units. Trials are needed to assess whether cryoprecipitate impacts clinical outcomes and to evaluate its safety.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fibrinogen from bovine plasma, Type I-S, 65-85% protein (≥75% of protein is clottable)
Human coagulation factor VIII concentrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fibrinogen from human plasma, 50-70% protein (≥80% of protein is clottable)
Sigma-Aldrich
Fibrinogen from human plasma, 35-65% protein (≥90% of protein is clottable).
Sigma-Aldrich
Fibrinogen from rat plasma, 60-80% protein (≥60% of protein is clottable)