Merck
Aliskiren limits abdominal aortic aneurysm, ventricular hypertrophy and atherosclerosis in an apolipoprotein-E-deficient mouse model.
Clinical Science (2014)
Seto SW, Krishna SM, Moran CS, et al.