Merck
  • The effect of cyclosporin-A on peri-operative myocardial injury in adult patients undergoing coronary artery bypass graft surgery: a randomised controlled clinical trial.
The effect of cyclosporin-A on peri-operative myocardial injury in adult patients undergoing coronary artery bypass graft surgery: a randomised controlled clinical trial.
Heart (British Cardiac Society) (2014-02-04)
Dj Hausenloy, G Kunst, E Boston-Griffiths, S Kolvekar, S Chaubey, L John, J Desai, Dm Yellon
ABSTRAKT
Cyclosporin-A (CsA) has been reported to reduce myocardial infarct size in both the experimental and clinical settings. This protective effect is dependent on its ability to prevent the opening of the mitochondrial permeability transition pore, a critical determinant of cell death in the setting of acute ischaemia-reperfusion injury. Whether CsA can reduce the extent of peri-operative myocardial injury (PMI) in patients undergoing coronary artery bypass graft (CABG) surgery is unknown, and is investigated in this randomised controlled clinical trial. 78 adult patients undergoing elective CABG surgery were randomised to receive either an intravenous bolus of CsA (2.5 mg/kg) or placebo administered after induction of anaesthesia and prior to sternotomy. PMI was assessed by measuring serum cardiac enzymes, troponin T (cTnT) and CK-MB at 0, 6, 12, 24, 48 and 72 h after surgery. There was no significant difference in mean peak cTnT levels between control (n=43) and CsA treatment (n=40) patients (0.56±0.06 ng/mL with control vs 0.35±0.05 ng/mL with CsA; p=0.07). However, in higher-risk patients with longer cardiopulmonary bypass times, there was a significant reduction in PMI with CsA therapy (p=0.049), with a reduced postoperative cTnT rise by 0.03 ng/mL for every 10 min, when compared with control. In patients with longer cardiopulmonary bypass times, a single intravenous bolus of CsA administered prior to CABG surgery reduced the extent of PMI.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, Molecular Biology, ≥95%
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Supelco
Cyclosporine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cyclosporin A solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Ciclosporin, European Pharmacopoeia (EP) Reference Standard