Merck
John Austin Milner, PhD (1947-2013).
The Journal of nutrition (2014-02-07)
Cindy D Davis, Regan L Bailey, Johanna T Dwyer, Paul M Coates
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Selenium, pellets, < 5mm, ≥99.999%