Merck
Buprenorphine-naloxone therapy in pain management.
Anesthesiology (2014-02-11)
Kelly Yan Chen, Lucy Chen, Jianren Mao
ABSTRAKT
Buprenorphine-naloxone (bup/nal in 4:1 ratio; Suboxone; Reckitt Benckiser Pharmaceuticals Incorporation, Richmond, VA) is approved by the Food and Drug Administration for outpatient office-based addiction treatment. In the past few years, bup/nal has been increasingly prescribed off-label for chronic pain management. The current data suggest that bup/nal may provide pain relief in patients with chronic pain with opioid dependence or addiction. However, the unique pharmacological profile of bup/nal confers it to be a weak analgesic that is unlikely to provide adequate pain relief for patients without opioid dependence or addiction. Possible mechanisms of pain relief by bup/nal therapy in opioid-dependent patients with chronic pain may include reversal of opioid-induced hyperalgesia and improvement in opioid tolerance and addiction. Additional studies are needed to assess the implication of bup/nal therapy in clinical anesthesia and perioperative pain management.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Naloxone hydrochloride dihydrate, ≥98% (TLC and titration), powder
Supelco
Buprenorphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Buprenorphine hydrochloride solution, analytical standard, for drug analysis
Naloxone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Naloxone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Buprenorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®