Merck
Continuous infusion of local anaesthetic following laparoscopic hysterectomy--a randomised controlled trial.
BJOG : an international journal of obstetrics and gynaecology (2014-02-20)
V Andrews, J T Wright, F Zakaria, S Banerjee, K Ballard
ABSTRAKT
To estimate whether a continuous infusion of intraperitoneal local anaesthetic for 48 hours following laparoscopic hysterectomy reduced the need for opioids delivered with a patient-controlled analgesia pump. Double-blind randomised placebo-controlled trial. District general hospital in the UK. Women undergoing a laparoscopic hysterectomy for a benign indication. Women were randomised to receive either 0.5% levobupivicaine or 0.9% normal saline via an ON-Q elastomeric pump for 48 hours postoperatively. The amount of opioids used via the patient-controlled analgesia pump was recorded and pain was measured using an 11-point Box Scale. The primary outcome was the amount of patient-administered morphine used over the first 48 postoperative hours. Secondary outcomes were length of hospital stay, oral analgesia use and level of patient-reported pain. Sixty women participated and completed the trial. There was no difference (P = 0.59) in the median amount of patient-administered morphine used between the levobupivicaine (23 mg) and placebo (18.5 mg) groups; median group difference 3.0 (95% CI -7.0 to 14.0). There was also no difference in the length of hospital stay with 40% of the treatment group remaining in hospital >48 hours compared with 30% of the placebo group (P = 0.08). Pain scores at all postoperative time points remained similar, with a median group difference in pain scores of 1.0 (95% CI -1.0 to 2.0) at the end of the first postoperative day. Continuous infusion of 0.5% levobupivicaine into the peritoneal cavity following laparoscopic hysterectomy does not have any opioid-sparing effects.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Bupivacaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Morphine sulfate salt solution, 1.0 mg/mL in methanol, drug standard
Morphine sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Morphine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Morphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®