Merck
Aspirin augments the expression of Adenomatous Polyposis Coli protein by suppression of IKKβ.
Biochemical and biophysical research communications (2014-03-13)
Noboru Ashida, Masako Kishihata, Dat Nguyen Tien, Kaeko Kamei, Takeshi Kimura, Masayuki Yokode
ABSTRAKT
Aspirin has been widely used as analgesic, antipyretic and anti-inflammatory medicine for long. In addition to these traditional effects, clinical studies suggest that aspirin can protect against cancer, but its mechanism has not been explored. To unveil it, we identified the proteins up- or down-regulated after incubation with aspirin by using proteomics analysis with Nano-flow LC/MALDI-TOF system. Interestingly, the analysis identified the protein of Adenomatous Polyposis Coli (APC) as one of the most up-regulated protein. APC regulates cell proliferation or angiogenesis, and is widely known as a tumor-suppressing gene which can cause colorectal cancer when it is mutated. Western blots confirmed this result, and real-time PCR indicated it is transcriptionally regulated. We further tried to elucidate the molecular mechanism with focusing on IKKβ. IKKβ is the essential kinase in activation of nuclear factor-kappa B (NF-κB), major transcriptional factors that regulate genes responsible for inflammation or immune response. Previous reports indicated that aspirin specifically inhibits IKKβ activity, and constitutively active form of IKKβ accelerates APC loss. We found that aspirin suppressed the expression of IKKβ, and the deletion of IKKβ by siRNA increases the expression of APC in HEK294 cells. Finally, we observed similar effects of aspirin in human umbilical vein endothelial cells. Taken together, these results reveal that aspirin up-regulates the expression of APC via the suppression of IKKβ. This can be a mechanism how aspirin prevents cancer at least in part, and a novel link between inflammatory NF-κB signaling and cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Aspiryna, United States Pharmacopeia (USP) Reference Standard
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard