Merck
  • High glucose-induced O-GlcNAcylated carbohydrate response element-binding protein (ChREBP) mediates mesangial cell lipogenesis and fibrosis: the possible role in the development of diabetic nephropathy.
High glucose-induced O-GlcNAcylated carbohydrate response element-binding protein (ChREBP) mediates mesangial cell lipogenesis and fibrosis: the possible role in the development of diabetic nephropathy.
The Journal of biological chemistry (2014-03-13)
Min-Jung Park, Dong-Il Kim, Seul-Ki Lim, Joo-Hee Choi, Ho-Jae Han, Kyung-Chul Yoon, Soo-Hyun Park
ABSTRAKT
Carbohydrate response element-binding protein (ChREBP) is a transcription factor responsible for carbohydrate metabolism in the liver. However, the role of ChREBP in diabetic nephropathy has not been elucidated. Thus, we investigated the role of ChREBP in mesangial cells in diabetic nephropathy. Treatment with 25 mM glucose (high glucose; HG) increased cellular O-GlcNAc and O-GlcNAcylated ChREBP in mesangial cells compared with normal 5.5 mM glucose. O-(2-acetamido-2-deoxy-D-glucopyranosylidene) amino N-phenylcarbamate (PUGNAc), a drug that increases O-GlcNAc, augmented the expression of ChREBP targets, whereas DON, a drug that decreases O-GlcNAc and O-GlcNAcase overexpression, mitigated the increase with HG. O-GlcNAc augmented the protein stability, transcriptional activity, and nuclear translocation of ChREBP. HG treatment also stimulated lipid accumulation and the contents of triglyceride and cholesterol in mesangial cells. In addition, HG triggered expression of hypoxia-inducible factor 1-α, vascular endothelial growth factor, and extracellular matrix components related to nephrosclerosis. The ChREBP mutant, W130A, did not exhibit HG-induced lipid accumulation and fibrotic proteins, suggesting that the Trp-130 residue in the MCR3 domain is important in the development of glomerulosclerosis. O-GlcNAcylated ChREBP was elevated in mesangium cells of streptozotocin-induced diabetic rats. In conclusion, HG increased the O-GlcNAcylated ChREBP level, which resulted in lipid accumulation and up-regulation of fibrotic proteins in mesangial cells. These effects may lead mesangial cells to an ultimately pathological state.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
N-Acetylglucosamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥95% (HPLC)
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥99% (HPLC)
Sigma-Aldrich
N-Acetyl-D-glucosamine, suitable for cell culture, BioReagent
Sigma-Aldrich
PUGNAc, ≥95% (HPLC)