Merck
Clinical significance of polypeptide N-acetylgalactosaminyl transferase-5 (GalNAc-T5) expression in patients with gastric cancer.
British journal of cancer (2014-03-13)
H He, Z Shen, H Zhang, X Wang, Z Tang, J Xu, Y Sun
ABSTRAKT
The family of polypeptide N-acetylgalactosaminyl transferases is responsible for the altered O-linked glycosylation occurring during the development of various cancers and their progression via altering O-glycan biosynthesis. Our studies were designed to investigate the expression and prognostic values of GalNAc-T5 and improve the risk stratification in patients with gastric cancer. Tissue samples from a training set and a validation set of patients with gastric adenocarcinoma from China were used for analyses. GalNAc-T5 expression was retrospectively analysed by immunohistochemistry. Results were assessed for association with clinicopathological parameters and overall survival by using Kaplan-Meier analysis. Prognostic values of GalNAc-T5 expression and clinical outcomes were evaluated by Cox regression analysis. A molecular prognostic stratification scheme incorporating GalNAc-T5 expression was determined by using receiver operating characteristic analysis. GalNAc-T5 expression was markedly reduced in gastric cancer tissues compared with non-malignant gastric mucosa. Low intratumoral GalNAc-T5 density, which was associated with tumour cell differentiation, T classification, N classification, and TNM stage in the two independent sets, was an independent prognosticator for poor prognosis of gastric cancer patients. Applying the prognostic value of intratumoral GalNAc-T5 density to the conventional clinicopathologic TNM stage system showed a better prognostic value in patients with gastric cancer. Intratumoral GalNAc-T5 expression was recognised as an independent prognostic marker for the overall survival of gastric cancer patients. Detection of GalNAc-T5 expression in gastric cancer tissues might add some prognostic information for patients with this disease and lead to a more accurate classification under the TNM stage system.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-GALNT5 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GALNT5 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Phosphorylase b from rabbit muscle, For use as a marker in SDS-PAGE
Sigma-Aldrich
Phosphorylase b from rabbit muscle, lyophilized powder, ≥20 units/mg protein, 2× crystallization