Merck
Thioredoxin-interacting protein promotes islet amyloid polypeptide expression through miR-124a and FoxA2.
The Journal of biological chemistry (2014-03-15)
Gu Jing, Clara Westwell-Roper, Junqin Chen, Guanlan Xu, C Bruce Verchere, Anath Shalev
ABSTRAKT
Thioredoxin-interacting protein (TXNIP) is up-regulated by glucose and diabetes and plays a critical role in glucotoxicity, inflammation, and beta-cell apoptosis, whereas we have found that TXNIP deficiency protects against diabetes. Interestingly, human islet amyloid polypeptide (IAPP) is also induced by glucose, aggregates into insoluble amyloid fibrils found in islets of most individuals with type 2 diabetes and promotes inflammation and beta-cell cytotoxicity. However, so far no connection between TXNIP and IAPP signaling had been reported. Using TXNIP gain and loss of function experiments, INS-1 beta-cells and beta-cell-specific Txnip knock-out mice, we now found that TXNIP regulates IAPP expression. Promoter analyses and chromatin-immunoprecipitation assays further demonstrated that TXNIP increases IAPP expression at the transcriptional level, and we discovered that TXNIP-induced FoxA2 (forkhead box A2) transcription factor expression was conferring this effect by promoting FoxA2 enrichment at the proximal FoxA2 site in the IAPP promoter. Moreover, we found that TXNIP down-regulates miR-124a expression, a microRNA known to directly target FoxA2. Indeed, miR-124a overexpression led to decreased FoxA2 expression and IAPP promoter occupancy and to a significant reduction in IAPP mRNA and protein expression and also effectively inhibited TXNIP-induced IAPP expression. Thus, our studies have identified a novel TXNIP/miR-124a/FoxA2/IAPP signaling cascade linking the critical beta-cell signaling pathways of TXNIP and IAPP and thereby provide new mechanistic insight into an important aspect of transcriptional regulation and beta-cell biology.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thioredoxin from Escherichia coli, recombinant, expressed in E. coli, essentially salt-free, lyophilized powder, ≥3 units/mg protein