Merck
  • Feasibility of the fludarabine busulfan 3 days and ATG 2 days reduced toxicity conditioning in 51 allogeneic hematopoietic stem cell transplantation: a single-center experience.
Feasibility of the fludarabine busulfan 3 days and ATG 2 days reduced toxicity conditioning in 51 allogeneic hematopoietic stem cell transplantation: a single-center experience.
Leukemia research (2014-03-25)
S P Chantepie, A C Gac, O Reman
ABSTRAKT
In reduced-toxicity conditioning hematopoietic stem cell transplantation, several studies failed to demonstrate the superiority of one conditioning over another. This study described 51 patients (median age of 58 years) allografted with the new FB3-ATG2 conditioning regimen for myeloid (66%) or lymphoid disease (33%). Comorbidity index ≥3 was noted in 23.5% of patients. Toxicities were close to those observed with RIC. One-year cumulative incidence of acute and chronic GVHD was 18.9% and 39.2%. The 2-year-NRM, DFS and OS were 25%, 57.5% and 66%. The FB3-ATG2 regimen is safe and efficient in both lymphoid and myeloid disorders. However, prospective comparative studies are needed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Fluoroadenine-9-β-D-arabinofuranoside, DNA synthesis and methylation inhibitor
Sigma-Aldrich
Adenine 9-β-D-arabinofuranoside, ≥99%
Busulfan, European Pharmacopoeia (EP) Reference Standard
Supelco
Busulfan, analytical standard, for drug analysis