Merck
Inhibitory effect of biosynthetic nanoscale peptide Melittin on hepatocellular carcinoma, driven by survivin promoter.
Journal of biomedical nanotechnology (2014-04-17)
Lili Qu, Min Jiang, Zhiyang Li, Fuxing Pu, Lailing Gong, Li Sun, Rujun Gong, Guozhong Ji, Jin Si
ABSTRAKT
Hepatic resection and orthotopic liver transplantation are the only potentially curative treatments for hepatocellular carcinoma (HCC) but are indicated only in a minority of patients. Biosynthetic nanoscale peptide Melittin (Mel) is postulated to disrupt microbial phospholipid membranes by formation of stable or transient pores. Survivin, a member of the inhibitor of apoptosis family, is transcriptionally upregulated in most malignant tissues but not in normal tissues. It has been reported that the survivin promoter activity is tumor-specific and makes it a good candidate for construction of gene therapy vectors. In the present study, a non-viral vector (pSURV-Mel), encoding Mel gene, was developed to evaluate its anti-tumor effect in HCC cell lines and in vivo in a mouse model of human HCC xenograft tumor. Our results showed that the survivin promoter is specifically activated in tumor cells, and the pSURV-Mel plasmid expressed Mel selectively in tumor cells and also induced cytotoxicity. Moreover, intratumoral Injection of pSURV-Mel significantly suppressed the growth of xenograft tumors. Mechanistically, pSURV-Mel induced cell death by an apoptosis-dependent pathway. All taken together, this study elucidates a relatively safe, highly effective and cancer specific gene therapy strategy for HCC. The mechanisms of non-viral vector-induced cell death which were revealed by this work will shed light on the construction of more powerful vectors for cancer therapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Melittin, ≥97% (HPLC), synthetic
Sigma-Aldrich
Melittin from honey bee venom, ≥85% (HPLC)
Sigma-Aldrich
Melittin from honey bee venom, ≥65% (HPLC)