Merck
Enhancement of an anti-tumor immune response by transient blockade of central T cell tolerance.
The Journal of experimental medicine (2014-04-23)
Imran S Khan, Maria L Mouchess, Meng-Lei Zhu, Bridget Conley, Kayla J Fasano, Yafei Hou, Lawrence Fong, Maureen A Su, Mark S Anderson
ABSTRAKT
Thymic central tolerance is a critical process that prevents autoimmunity but also presents a challenge to the generation of anti-tumor immune responses. Medullary thymic epithelial cells (mTECs) eliminate self-reactive T cells by displaying a diverse repertoire of tissue-specific antigens (TSAs) that are also shared by tumors. Therefore, while protecting against autoimmunity, mTECs simultaneously limit the generation of tumor-specific effector T cells by expressing tumor self-antigens. This ectopic expression of TSAs largely depends on autoimmune regulator (Aire), which is expressed in mature mTECs. Thus, therapies to deplete Aire-expressing mTECs represent an attractive strategy to increase the pool of tumor-specific effector T cells. Recent work has implicated the TNF family members RANK and RANK-Ligand (RANKL) in the development of Aire-expressing mTECs. We show that in vivo RANKL blockade selectively and transiently depletes Aire and TSA expression in the thymus to create a window of defective negative selection. Furthermore, we demonstrate that RANKL blockade can rescue melanoma-specific T cells from thymic deletion and that persistence of these tumor-specific effector T cells promoted increased host survival in response to tumor challenge. These results indicate that modulating central tolerance through RANKL can alter thymic output and potentially provide therapeutic benefit by enhancing anti-tumor immunity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC), suitable for fluorescence
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, suitable for fluorescence, BioReagent, ≥95.0% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining