Merck
Two plants to put 'clean coal' to test.
Nature (2014-05-03)
Richard Van Noorden
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C