Merck
A form of the metabolic syndrome associated with mutations in DYRK1B.
The New England journal of medicine (2014-05-16)
Ali R Keramati, Mohsen Fathzadeh, Gwang-Woong Go, Rajvir Singh, Murim Choi, Saeed Faramarzi, Shrikant Mane, Mohammad Kasaei, Kazem Sarajzadeh-Fard, John Hwa, Kenneth K Kidd, Mohammad A Babaee Bigi, Reza Malekzadeh, Adallat Hosseinian, Masoud Babaei, Richard P Lifton, Arya Mani
ABSTRAKT
Genetic analysis has been successful in identifying causative mutations for individual cardiovascular risk factors. Success has been more limited in mapping susceptibility genes for clusters of cardiovascular risk traits, such as those in the metabolic syndrome. We identified three large families with coinheritance of early-onset coronary artery disease, central obesity, hypertension, and diabetes. We used linkage analysis and whole-exome sequencing to identify the disease-causing gene. A founder mutation was identified in DYRK1B, substituting cysteine for arginine at position 102 in the highly conserved kinase-like domain. The mutation precisely cosegregated with the clinical syndrome in all the affected family members and was absent in unaffected family members and unrelated controls. Functional characterization of the disease gene revealed that nonmutant protein encoded by DYRK1B inhibits the SHH (sonic hedgehog) and Wnt signaling pathways and consequently enhances adipogenesis. Furthermore, DYRK1B promoted the expression of the key gluconeogenic enzyme glucose-6-phosphatase. The R102C allele showed gain-of-function activities by potentiating these effects. A second mutation, substituting proline for histidine 90, was found to cosegregate with a similar clinical syndrome in an ethnically distinct family. These findings indicate a role for DYRK1B in adipogenesis and glucose homeostasis and associate its altered function with an inherited form of the metabolic syndrome. (Funded by the National Institutes of Health.).