Merck
Patent foramen ovale (PFO) closure to prevent recurrent stroke in a patient with a nickel allergy.
Connecticut medicine (2014-05-17)
Asad A Rizvi, Ronan Margey, Francis Kiernan
ABSTRAKT
The case of a 44-year-old female with multiple embolic cerebellar infarcts associated with a "high risk" patent foramen ovale (PFO) is presented. This article reviews current management of PFO in association with cryptogenic stroke in light of the results of three recent randomized controlled trials and two meta-analyses of observational data. The article will also discuss circumstances which merit consideration for closure despite the negative trial data, and will review our management strategy for closure in this patient with a nickel allergy that precluded the use of the most commonly used PFO closure device.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 4200, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 3202, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Nickel, powder, <1 μm, 99.8% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Nickel, foil, 15mm disks, thickness 0.005mm, as rolled, 99.9%
Nickel, tube, 100mm, outside diameter 1.8mm, inside diameter 1.6mm, wall thickness 0.1mm, hard, 99.5%
Nickel, tube, 200mm, outside diameter 0.8mm, inside diameter 0.5mm, wall thickness 0.15mm, as drawn, 99.5%
Nickel, tube, 1000mm, outside diameter 2.5mm, inside diameter 2.25mm, wall thickness 0.125mm, hard, 99.5%
Nickel, wire reel, 200m, diameter 0.15mm, as drawn, 99%
Nickel, foil, 10mm disks, thickness 0.125mm, annealed, 99%
Nickel, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%
Nickel, foil, 1m coil, thickness 0.50mm, annealed, 99%
Nickel, foil, 1m coil, thickness 0.05mm, as rolled, 99.98%
Nickel, foil, 15mm disks, thickness 0.05mm, as rolled, 99.99+%
Nickel, tube, 100mm, outside diameter 7.11mm, inside diameter 6.57mm, wall thickness 0.27mm, hard, 99.5%
Nickel, foil, 15mm disks, thickness 0.25mm, annealed, 99%
Nickel, foil, 1m coil, thickness 0.20mm, coil width 1mm, annealed, 99%
Nickel, foil, 10mm disks, thickness 0.125mm, annealed, 99.98%
Nickel, wire reel, 0.2m, diameter 0.4mm, as drawn, 99.99+%
Nickel, foil, 10mm disks, thickness 0.0025mm, 99.95%
Nickel, foil, 0.5m coil, thickness 0.015mm, 99.9%
Nickel, wire reel, 10m, diameter 0.5mm, hard, 99.98%
Nickel, foil, 150x150mm, thickness 0.30mm, as rolled, 99.98%
Nickel, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99+%