Merck
Susceptibility to chlorpyrifos in pyrethroid-resistant populations of Aedes aegypti (Diptera: Culicidae) from Mexico.
Journal of medical entomology (2014-06-06)
Beatriz Lopez, Gustavo Ponce, Jessica A Gonzalez, Selene M Gutierrez, Olga K Villanueva, Gabriela Gonzalez, Cristina Bobadilla, Iram P Rodriguez, William C Black, Adriana E Flores
ABSTRAKT
Resistance to the organophosphate insecticide chlorpyrifos was evaluated in females from six strains of Aedes aegypti (L.) that expressed high levels of cross-resistance to eight pyrethroid insecticides. Relative to LC50 and LC90 at 24 h of a susceptible New Orleans (NO) strain, three strains were highly resistant to chlorpyrifos (Coatzacoalcos, resistance ratio [RRLC90 = 11.97; Pozarica, RRLC90 = 12.98; and Cosoleacaque, RRLC50 = 13.94 and RRLC90 = 17.57), one strain was moderately resistant (Veracruz, RRLC90 = 5.92), and two strains were susceptible (Tantoyuca and Martinez de la Torre, RRLC50 and RRLC90 < 5) in bottle bioassays according to Centers for Disease Control and Prevention. Furthermore, high levels of alpha- or beta-esterase activity in the sample populations were correlated with resistance, suggesting that esterase activity may be a mechanism causing the development of organophosphate resistance in these populations. Overall, the populations in this study were less resistant to chlorpyrifos than to pyrethroids. Rotation of insecticides used in control activities is recommended to delay or minimize the occurrence of high levels of resistance to chlorpyrifos among local populations of Ae. aegypti. The diagnostic dose and diagnostic time for chlorpyrifos resistance monitoring was determined to be 85 microg per bottle and 30 min, respectively, using the susceptible NO strain.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Chlorpyrifos, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Chlorpyrifos, PESTANAL®, analytical standard