Merck
Remote sensing. Carbon-mapping satellite will monitor plants' faint glow.
Science (New York, N.Y.) (2014-06-14)
Eric Hand
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Millipore
Activated charcoal, suitable for GC
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Carbon, rod, 100mm, diameter 13.0mm, graphite, 100%
Carbon, rod, 25mm, diameter 50mm, graphite, 99.95%
Carbon, foil, 8mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.2mm, pyrolytic graphite, 99.99%
Carbon, foil, 50x50mm, thickness 0.25mm, rigid graphite, fine grain size, 99.997%
Carbon, foil, 10x10mm, thickness 2.0mm, hOpg
Carbon, microleaf, 25x70mm, thinness 0.025μm, specific density 5μg/cm2, temporary glass support, 99.997%
Carbon, microleaf, 25x70mm, thinness 0.25μm, specific density 50μg/cm2, temporary glass support, 99.997%
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Carbon, foil, 6mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, rod, 300mm, diameter 13.0mm, graphite, 100%
Carbon, rod, 100mm, diameter 2.0mm, graphite, 100%
Carbon, foil, not light tested, 100x100mm, thickness 0.075mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon, foil, 5x10mm, thickness 2.0mm, hOpg