- Synthesis of an array of potential matrix metalloproteinase inhibitors using a sequence of polymer-supported reagents.
Synthesis of an array of potential matrix metalloproteinase inhibitors using a sequence of polymer-supported reagents.
Bioorganic & medicinal chemistry letters (1999-08-18)
M Caldarelli, J Habermann, S V Ley
PMID10450979
ABSTRAKT
Polymer-supported reagents and sequestering agents may be used to generate an array of variously substituted hydroxamic acid derivatives as potential inhibitors of matrix metalloproteinases without any chromatographic purification step.