Merck
ART-27, an androgen receptor coactivator regulated in prostate development and cancer.
The Journal of biological chemistry (2004-01-09)
Samir S Taneja, Susan Ha, Nicole K Swenson, Inés Pineda Torra, Serge Rome, Paul D Walden, Hong Ying Huang, Ellen Shapiro, Michael J Garabedian, Susan K Logan
ABSTRAKT
Androgen receptor trapped clone-27 (ART-27) is a newly described transcriptional coactivator that binds to the N terminus of the androgen receptor (AR). Given the vital importance of AR signaling in prostate growth and differentiation, we investigated the role of ART-27 in these processes. Immunohistochemical studies indicate that ART-27 protein is expressed in differentiated epithelial cells of adult human prostate and breast tissue. In prostate, ART-27 is abundant in AR-positive prostate luminal epithelial cells, in contrast to the stroma, where cells express AR but not ART-27. The use of a rat model of androgen depletion/reconstitution indicates that ART-27 expression is associated with the elaboration of differentiated prostate epithelial cells. Interestingly, regulated expression of ART-27 in the androgen-sensitive LNCaP prostate cancer cell line inhibits androgen-mediated cellular proliferation and enhances androgen-mediated transcription of the prostate-specific antigen (PSA) gene. Consistent with a growth suppressive function, we show that ART-27 expression levels are negligible in human prostate cancer. Importantly, examination of ART-27 protein expression in early fetal prostate development demonstrates that ART-27 is detected only when the developing prostate gland has proceeded from a solid mass of undifferentiated cells to a stage in which differentiated luminal epithelial cells are evident. Thus, ART-27 is an AR cofactor shown to be subject to both cell type and developmental regulation in humans. Overall, the results suggest that decreased levels of ART-27 protein in prostate cancer tissue may occur as a result of de-differentiation, and indicate that ART-27 is likely to regulate a subset of AR-responsive genes important to prostate growth suppression and differentiation.