Merck
Anti-cytokeratin 5/6: a positive marker for epithelioid mesothelioma.
Histopathology (1997-08-01)
J Clover, J Oates, C Edwards
ABSTRAKT
Previous studies using frozen material have suggested that cytokeratin 5 is expressed by pleural mesothelioma but not by adenocarcinoma. In the present study, reactions for cytokeratins 5 and 6 were investigated in 33 pleural mesotheliomas and 27 secondary adenocarcinomas of the pleura using formalin-fixed, paraffin-embedded material and a commercially available antibody (anti-cytokeratin 5/6). All of the adenocarcinomas originated in the peripheral lung. The epithelioid component of the mesotheliomas gave a strongly positive reaction: the reaction in sarcomatoid or desmoplastic areas was absent or weak. Twenty-two of the adenocarcinomas were negative, in four there was a weak, equivocal reaction, and in one there was patchy positivity. We conclude that this antibody is potentially a useful positive marker for the identification of the epithelioid variant of mesothelioma in formalin-fixed and paraffin-embedded material.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Cytokeratin 5 Antibody, 6, clone D5/16B4, clone D5/16B4, Chemicon®, from mouse