Merck
The nuclear vitamin D receptor: biological and molecular regulatory properties revealed.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research (1998-04-03)
M R Haussler, G K Whitfield, C A Haussler, J C Hsieh, P D Thompson, S H Selznick, C E Dominguez, P W Jurutka