Merck
In vivo studies on butylated hydroxyanisole.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (1999-10-29)
F Iverson
MATERIAŁY
Numer produktu
Marka
Opis produktu
Butylhydroxyanisole, European Pharmacopoeia (EP) Reference Standard
Supelco
Butylated hydroxyanisole, analytical standard
Sigma-Aldrich
Butylated hydroxyanisole, 99%, FCC, FG
Sigma-Aldrich
Butylated hydroxyanisole, ≥98.5%
Supelco
Butylated hydroxyanisole, Pharmaceutical Secondary Standard; Certified Reference Material