Merck
[Vanillylmandelic acid (VMA) and homovanillic acid (HVA)].
Nihon rinsho. Japanese journal of clinical medicine (2001-02-07)
Y Kanesaka, K Tomita
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent