Merck
Two cases of confluent and reticulate papillomatosis: successful treatments of one case with cefdinir and another with minocycline.
The Journal of dermatology (2000-10-29)
A Yamamoto, Y Okubo, H Oshima, T Oh-i, M Koga
ABSTRAKT
The present report presents two cases of confluent and reticulate papillomatosis. Case 1 was a 24-year-old man who had suffered from skin eruptions for six months, and Case 2 was a 19-year-old woman who had had this disease for three days. In both patients, reticular dark brown papules, accompanied by mild keratosis and infiltration, spread from the trunk to the neck and upper arm. Direct light microscopy did not detect the presence of any fungi, and histopathological examinations confirmed hyperkeratosis, acanthosis, papillomatosis, and mild small-round-cell infiltration. Thus, these patients were diagnosed as confluent and reticulate papillomatosis. Neither one had diabetes or thyroid dysfunction. In Case 1, cefdinir was effective, and in Case 2, minocycline hydrochloride and ketoconazole were effective. To the best of our knowledge, this was the first documented case of confluent and reticulate papillomatosis responding to cefdinir.