Merck
Gamma-hydroxybutyrate, gamma-butyrolactone, and 1,4-butanediol: a case report and review of the literature.
Pediatric emergency care (2001-01-04)
M Shannon, L S Quang
ABSTRAKT
GHB, GBL, and 1,4-BD are prevalent drugs of abuse in the United States. Unfortunately, attempts to regulate GHB have been circumvented by clandestine trafficking through the Internet and marketing of "natural" chemical precursors . Despite repeated FDA warnings to the public about their dangers as well as recent federal scheduling of GHB and GBL, they remain accessible as "club drugs" on Internet websites, as natural dietary supplements in health food stores, and as illicit products manufactured at home or in clandestine laboratories. EDs and poison control centers nationwide will undoubtedly continue to manage GHB, GBL, and 1,4-BD toxicities.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
γ-Butyrolactone, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Hydroxybutanoic acid lactone, ≥98%, FCC, FG
Supelco
γ-Butyrolactone, analytical standard