Merck
Codeine phosphate in paediatric medicine.
British journal of anaesthesia (2001-09-28)
D G Williams, D J Hatch, R F Howard
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Codeine solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Codeine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Codeine, European Pharmacopoeia (EP) Reference Standard