Merck
Myth: silver sulfadiazine is the best treatment for minor burns.
The Western journal of medicine (2001-08-31)
J Y Chung, M E Herbert
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Sulfadiazyna srebra, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Silver(I) sulfadiazine, 98%