Merck
Antidote review: fomepizole for methanol poisoning.
American journal of therapeutics (2003-01-11)
Mark B Mycyk, Jerrold B Leikin
ABSTRAKT
Fomepizole (Antizol) was recently approved by the US Food and Drug Administration for treatment of methanol poisoning. By inhibiting the hepatic enzyme alcohol dehydrogenase, it presents formation of toxic metabolites with far fewer consequences than traditional ethanol therapy. It appears that fomepizole will become standard therapy for methanol intoxication as it is for ethylene glycol poisoning.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fomepizole, 99%