Merck
Critical care management of major hydrofluoric acid burns: a case report, review of the literature, and recommendations for therapy.
Burns : journal of the International Society for Burn Injuries (2004-05-18)
Martin W Dünser, Markus Ohlbauer, Josef Rieder, Isabella Zimmermann, Helmut Ruatti, Anton H Schwabegger, Florian Bodrogi, Georg M Huemer, Barbara E Friesenecker, Andreas J Mayr, Philipp Lirk
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis