Merck
Anabolism and mechanism of action of Ro24-5098, an isomer of 2',3'-dideoxyadenosine (ddA) with anti-HIV activity.
Annals of the New York Academy of Sciences (1990-01-01)
K B Frank, E V Connell, M J Holman, D M Huryn, B C Sluboski, S Y Tam, L J Todaro, M Weigele, D D Richman, H Mitsuya
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2′,3′-Dideoxyadenosine, ≥97% (HPLC)
Didanosine impurity G, European Pharmacopoeia (EP) Reference Standard