Merck
Allergy to formaldehyde and ethylene-oxide.
Clinical reviews in allergy (1991-01-01)
J Bousquet, F B Michel
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylene oxide solution, 2.5-3.3 M in THF
Supelco
Ethylene oxide solution, certified reference material, 50 mg/mL in methanol