Merck
[Study advance on formation, transformation and detection of masked deoxynivalenol].
Wei sheng yan jiu = Journal of hygiene research (2009-05-05)
Chuanchuan Yu, Hongxia Yu, Fengqin Li
ABSTRAKT
Conjugated deoxynivalenol, in which the toxins were usually bound to a more polar substance like glucose in crops, is referred to as masked deoxynivalenol. It could not detected with routine detection methods, but could release its toxic precursor after hydrolysis in gastrointestinal tract. In this paper, summarizes the formation, transformation and detection of masked deoxynivalenol as well as its relationship with human health were reviewed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Deoxynivalenol
Supelco
Deoxynivalenol solution, 100 μg/mL in acetonitrile, analytical standard
4-Deoxynivalenol in acetonitrile, IRMM®, certified reference material