Merck
Marine toxins and the cytoskeleton: okadaic acid and dinophysistoxins.
The FEBS journal (2008-11-20)
Carmen Vale, Luis M Botana
ABSTRAKT
Okadaic acid (OA) and its analogs, the dinophysistoxins, are potent inhibitors of protein phosphatases 1 and 2A. This action is well known to cause diarrhea and gastrointestinal symptons when the toxins reach the digestive tract by ingestion of mollusks. A less well-known effect of these group of toxins is their effect in the cytoskeleton. OA has been shown to stimulate cell motility, loss of stabilization of focal adhesions and a consequent loss of cytoskeletal organization due to an alteration in the tyrosine-phosphorylated state of the focal adhesion kinases and paxillin. OA causes cell rounding and loss of barrier properties through mechanisms that probably involve disruption of filamentous actin (F-actin) and/or hyperphosphorylation and activation of kinases that stimulate tight junction disassembly. Neither methyl okadaate (a weak phosphatase inhibitor) nor OA modify the total amount of F-actin, but both toxins cause similar changes in the F-actin cytoskeleton, with strong retraction and rounding, and in many cases cell detachment. OA and dinophysistoxin-1 (35S-methylokadaic acid) cause rapid changes in the structural organization of intermediate filaments, followed by a loss of microtubules, solubilization of intermediate filament proteins, and disruption of desmosomes. The detailed pathways that coordinate all these effects are not yet known.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Okadaic acid from Prorocentrum concavum, 92-100% (HPLC)