Merck
Cytotoxic and anticancer activities of isatin and its derivatives: a comprehensive review from 2000-2008.
Anti-cancer agents in medicinal chemistry (2009-05-16)
K L Vine, L Matesic, J M Locke, M Ranson, D Skropeta
ABSTRAKT
Isatin (1H-indole-2,3-dione) and its derivatives demonstrate a diverse array of biological and pharmacological activities including anticonvulsant, antibacterial, antifungal, antiviral and anticancer properties. This broad spectrum of biochemical targets has been facilitated by the synthetic versatility of isatin, which has allowed the generation of a large number of structurally diverse derivatives including analogues derived from substitution of the aryl ring, and/or derivatisation of the isatin nitrogen and C2/C3 carbonyl moieties. The recent FDA approval of the oxindole sunitinib malate, as a kinase inhibitor for the treatment of advanced renal carcinoma and gastrointestinal stromal tumours, underscores the increasing interest in isatins as a new class of antineoplastic agents. In addition to potent kinase inhibition, the mechanism of action of other isatin derivatives includes the inhibition and/or modulation of proteases, translation initiation, neo-vascularisation and tubulin polymerisation. It was therefore the objective of this review to systematically evaluate the cytotoxic and anticancer properties of various substituted isatins and collate these findings to be used as a guide for future structure-activity relationship and mode of action studies. This is the first review to comprehensively discuss the in vitro and in vivo anticancer activities of isatin and its substituted derivatives.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Isatin, 97%