Merck
Vinyl chloride and the liver.
Journal of hepatology (2009-10-20)
Morris Sherman
ABSTRAKT
Vinyl chloride monomer is a known cause of angiosarcoma of the liver. It also has other toxic effects on the liver, and it has recently been suggested that exposure to vinyl chloride also causes hepatocellular carcinoma. However, the data on which this conclusion is based is incomplete. There is inadequate ascertainment of unequivocal diagnoses. In the largest studies lack of data meant that confounding diseases such as viral hepatitis or alcoholic liver disease could not be assessed. At best, the increase in risk is minimal, based on more than 22,000 exposed workers and more than 640,000 person years of observation. However, based on the available data the hypothesis that vinyl chloride causes or contributes to the development of hepatocellular carcinoma remains unproven.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Vinyl chloride solution, 200 μg/mL in methanol, analytical standard
Supelco
Vinyl chloride solution, certified reference material, 2000 μg/mL in methanol