Merck
Inhaled albuterol/salbutamol and ipratropium bromide and their combination in the treatment of chronic obstructive pulmonary disease.
Expert opinion on drug metabolism & toxicology (2010-02-19)
Joshiah Gordon, Ralph J Panos
ABSTRAKT
Chronic obstructive pulmonary disease (COPD) is a major cause of morbidity and mortality throughout the world. Combination therapy with albuterol and ipratropium bromide was approved > 15 years ago for the treatment of COPD. We review the mechanism of action, clinical efficacy, and safety of albuterol, ipratropium and combined albuterol-ipratropium therapy. We conducted a PubMed literature search using the keywords COPD, albuterol, ipratropium bromide and Combivent (Boehringer Ingelheim Corp., Ridgefield, CT, USA); pertinent references within the identified citations are included in the review. Data from the manufacturers are also evaluated. At the time of its approval, albuterol/ipratropium bromide was an innovative combination of existing medications for the treatment of COPD. The combined formulation provides better improvement in airflow than either component alone and, by reducing the number of separate inhalers, simplifies therapy and improves compliance compared with the individual components. The recent development and approval of longer acting and more potent beta agonists, anticholinergics and newer combination treatments have surpassed many of the advantages of combined albuterol-ipratropium for the treatment of patients with stable COPD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Salbutamol
Supelco
Salbutamol, VETRANAL®, analytical standard
Salbutamol, European Pharmacopoeia (EP) Reference Standard