Merck
[Mechanisms of antistress and antidepressive effects of magnesium and pyridoxine].
Zhurnal nevrologii i psikhiatrii imeni S.S. Korsakova (2010-02-03)
I Iu Torshin, O A Gromova, E I Gusev
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pyridoxine, ≥98%