Merck
Cyclosporin A--a review on fermentative production, downstream processing and pharmacological applications.
Biotechnology advances (2011-03-31)
Shrikant A Survase, Lalit D Kagliwal, Uday S Annapure, Rekha S Singhal
ABSTRAKT
In present times, the immunosuppressants have gained considerable importance in the world market. Cyclosporin A (CyA) is a cyclic undecapeptide with a variety of biological activities including immunosuppressive, anti-inflammatory, antifungal and antiparasitic properties. CyA is produced by various types of fermentation techniques using Tolypocladium inflatum. In the present review, we discuss the biosynthetic pathway, fermentative production, downstream processing and pharmacological activities of CyA.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, Molecular Biology, ≥95%
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Supelco
Cyclosporine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cyclosporin A solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Ciclosporin, European Pharmacopoeia (EP) Reference Standard