Merck
Retinoic acid generates regulatory T cells in experimental transplantation.
Transplantation proceedings (2011-08-16)
C Moore, C Fuentes, D Sauma, J Morales, M R Bono, M Rosemblatt, J A Fierro
ABSTRAKT
Regulatory T cells play a key role to inhibit effector lymphocytes, avoid, autoimmunity, and restrain allogeneic immunity. Retinoic acid is an important cofactor that stimulates the generation and expansion of regulatory T cells. Naive T cells, coincubated with allogeneic antigen-presenting cells and retinoic acid, in conjunction with transforming growth factor (TGF) β and interleukin (IL) 2, generated allogeneic regulatory T cells de novo. These cells were able to inhibit skin rejection in adoptive transfer experiments. The generation of regulatory T cells ex vivo with retinoic acid, TGF-β, and IL-2 represents a new step toward specific regulation of allogeneic immune responses.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Tretinoin, European Pharmacopoeia (EP) Reference Standard
USP
Tretynoina, United States Pharmacopeia (USP) Reference Standard
Supelco
Tretinoin, Pharmaceutical Secondary Standard; Certified Reference Material