Merck
Endomyocardial fibrosis--the possible connexion with myocardial levels of magnesium and cerium.
International journal of cardiology (1990-07-01)
S M Valiathan, C C Kartha
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cerium, chips
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Cerium, rod, 100mm, diameter 12.7mm, cast, 99.9%
Cerium, rod, 100mm, diameter 6.35mm, cast, 99.9%
Cerium, rod, 50mm, diameter 12.7mm, cast, 99.9%
Cerium, rod, 50mm, diameter 6.35mm, cast, 99.9%