Merck
Diethyltoluamide (DEET) insect repellent toxicosis.
The Veterinary clinics of North America. Small animal practice (1990-03-01)
D C Dorman
ABSTRAKT
Poisonings resulting from DEET are rarely encountered in companion animals. In human patients, DEET toxicosis has been associated with the development of a toxic encephalopathy characterized by tremors, seizures, behavioral changes, and abnormal movements. Generally, DEET toxicosis in companion animals is of short duration and is characterized by vomiting, tremors, ataxia, and excitation. Treatment for DEET toxicosis includes symptomatic therapy and decontamination.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N,N-Diethyl-3-methylbenzamide, 97%
Supelco
DEET, PESTANAL®, analytical standard