Merck
[The effect of translation inhibitor cycloheximide on expression of mammalian genes].
Biokhimiia (Moscow, Russia) (1990-07-01)
V N Chesnokov, N P Mertvetsov
ABSTRAKT
The data concerning the cycloheximide influence on individual mRNA content, transcription rate and chromatin structure in mammalian cells are reviewed. The putative mechanism of cycloheximide effect is explained as being due to the existence of short-living regulatory proteins. It is concluded that cycloheximide is a helpful tool in the study of regulation of gene expression in mammalian cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology