Merck
Carvedilol: a third-generation β-blocker should be a first-choice β-blocker.
Expert review of cardiovascular therapy (2011-12-14)
James J DiNicolantonio, Daniel G Hackam
ABSTRAKT
β-Blockers are a standard of care in many clinical settings such as acute myocardial infarction, heart failure and patients at risk for a coronary event. However, not all β-blockers are the same and they vary in properties such as lipophilicity, metabolic profile, receptor inhibition, hemodynamics, tolerability and antioxidant/anti-inflammatory effects. It has been unclear whether these differences affect outcomes or if one β-blocker should be preferred over another. This review will summarize the properties of metoprolol, atenolol and carvedilol, as well as comparative experimental and clinical trials between these agents. We will provide compelling evidence of why carvedilol should be a first-line β-blocker and why it offers many advantages over the β1-selective β-blockers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Carvedilol, ≥98% (HPLC), solid
Carvedilol, European Pharmacopoeia (EP) Reference Standard
Carvedilol for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Carvedilol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Carvedilol, United States Pharmacopeia (USP) Reference Standard