Merck
Edge loading has a paradoxical effect on wear in metal-on-polyethylene total hip arthroplasties.
Clinical orthopaedics and related research (2012-05-31)
William H Harris
ABSTRAKT
Edge wear is an adverse factor that can negatively impact certain THAs. In some metal-on-metal THAs, it can lead to adverse tissue reactions including aseptic lymphocytic vasculitis-associated lesions and even to pseudotumor formation. In some ceramic-on-ceramic THAs, it can lead to squeaking and/or stripe wear. Edge wear in metal-on-metal and ceramic-on-ceramic THAs can also be associated with accelerated wear across the articulation of these joints. I asked: Does edge wear occur in metal-on-polyethylene (MOP) articulations? And if so, does it increase joint wear? I examined the evidence in the literature for edge wear occurring in MOP THA and then assessed the evidence in the literature for data supporting the concept that edge wear in MOP hips could accelerate wear across the articulation over time. Extensive data in the literature confirm edge wear is common in MOP THA. Surprisingly, the evidence does not support that it accelerates wear across the articulation. In fact, substantial data support the concept that it does not. These observations suggest, in terms of edge wear accelerating overall wear, MOP articulation may have a privileged position compared to hard-on-hard THA articulations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Supelco
Polyethylene, analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Polyethylene (LDPE), ERM®, certified reference material