- No allergic reaction after TKA in a chrome-cobalt-nickel-sensitive patient: case report and review of the literature.
No allergic reaction after TKA in a chrome-cobalt-nickel-sensitive patient: case report and review of the literature.
Knee surgery, sports traumatology, arthroscopy : official journal of the ESSKA (2012-04-11)
Emmanuel Thienpont, Yorick Berger
PMID22488014
ABSTRAKT
Hypersensitivity to metallic implants remains relatively unpredictable and poorly understood. Although 20-25 % of total joint arthroplasty patients develop metal sensitivity, only a few highly susceptible persons (<1 %) exhibit symptoms. We present a case report of a fifty-two-year-old woman with a preoperatively documented metal allergy who underwent bilateral total knee arthroplasty using a titanium-niobium-coated implant on one side and a chrome-cobalt implant on the other side because of a logistics problem. At 2-year follow-up, no clinical symptoms of allergy or loosening of the implant were observed. Level of evidence IV.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Cobalt, foil, not light tested, 50x50mm, thickness 0.025mm, 99.9%
Cobalt, wire reel, 0.5m, diameter 0.5mm, hard, 99.99+%
Cobalt, wire reel, 2m, diameter 0.05mm, hard, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.008mm, 99.9%
Cobalt, wire reel, 0.2m, diameter 0.5mm, hard, 99.99+%
Cobalt, foil, 10mm disks, thickness 0.009mm, 99.9%
Cobalt, foil, 15mm disks, thickness 0.015mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.025mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.007mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Cobalt, foil, 6mm disks, thickness 0.006mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.005mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.125mm, annealed, 99.9%
Cobalt, foil, 4mm disks, thickness 0.025mm, 99.9%
Cobalt, microfoil, disks, 10mm, thinness 0.25μm, specific density 222μg/cm2, permanent mylar 3.5μm support, 99.9%
Cobalt, foil, 6mm disks, thickness 0.007mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.25mm, as rolled, 99.9%
Cobalt, foil, 8mm disks, thickness 0.05mm, as rolled, 99.9%