Merck
Clinical and histological features of idiosyncratic acute liver injury caused by temozolomide.
Digestive diseases and sciences (2012-12-06)
Lafaine M Grant, David E Kleiner, Hari S Conjeevaram, Raj Vuppalanchi, William M Lee
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Temozolomide, ≥98% (HPLC)
Supelco
Temozolomide, VETRANAL®, analytical standard