Merck
Metal release from hip prostheses: cobalt and chromium toxicity and the role of the clinical laboratory.
Clinical chemistry and laboratory medicine (2012-10-02)
Jonathon R Campbell, Mathew P Estey
ABSTRAKT
Individuals with advanced hip disease suffer from pain, impaired hip function, and decreased quality of life. Roughly one million metal-on-metal (MoM) hip prostheses have been implanted worldwide in order to ameliorate these issues. While most MoM hip replacements are successful, some patients suffer from serious adverse effects secondary to the release of metal debris due to implant wear and corrosion. MoM hip prostheses are comprised predominantly of cobalt and chromium, and the serum concentration of these metal ions has been shown to correlate with both implant wear and the accumulation of metal debris in the periprosthetic tissue. Consequently, measurement of cobalt and chromium concentrations may be useful in the assessment of implant function and the potential for adverse effects in the follow-up of patients with MoM hip prostheses. The purpose of this Mini Review is to describe the adverse biological consequences of metal release from hip prostheses, provide an overview of the clinical utility of cobalt and chromium measurement and the current recommendations for testing, and alert laboratorians and physicians to the many challenges associated with measuring these metal ions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Cobalt, foil, not light tested, 50x50mm, thickness 0.025mm, 99.9%
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Cobalt, wire reel, 0.5m, diameter 0.5mm, hard, 99.99+%
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Chromium, rod, 10mm, diameter 5.0mm, 99.7+%
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Cobalt, wire reel, 2m, diameter 0.05mm, hard, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.008mm, 99.9%
Chromium, rod, 25mm, diameter 2.0mm, 99.7+%
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Cobalt, wire reel, 0.2m, diameter 0.5mm, hard, 99.99+%
Cobalt, foil, 10mm disks, thickness 0.009mm, 99.9%
Cobalt, foil, 15mm disks, thickness 0.015mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.025mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.007mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Cobalt, foil, 6mm disks, thickness 0.006mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.005mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.9%